Learn More
AdipoGen Anhydrotetracycline hydrochloride,Chemical. CAS: 13803-65-1
SDP
Supplier:  AdipoGen CDXA0197GG25
Encompass_Preferred
Anhydrotetracycline hydrochloride, AdipoGen Life Sciences. Chemical. CAS: 13803-65-1. Formula: C22H22N2O7 . HCl. MW: 462.88. Synthetic. A tetracycline derivative exhibiting no antibiotic activity. A useful effector of tetracycline-dependent gene expression in bacterial expression systems.
Catalog No. 50-595-758