Learn More
Cayman Chemical Anhydrotetracycline hydrochl
SDP
Supplier:  Cayman Chemical 10009542250 MG
Encompass_Preferred
A powerful effector in both the TetR and revTetR transcriptional regulator systems, binding the Tet repressor 35-fold more strongly than Tet; does not act a general inhibitor of translation and is a poor antibiotic
Catalog No. 50-288-8003