Learn More
Cayman Chemical Anhydrotetracycline hydrochl
SDP
Catalog No. 502890219
Encompass_Preferred
A powerful effector in both the TetR and revTetR transcriptional regulator systems, binding the Tet repressor 35-fold more strongly than Tet; does not act a general inhibitor of translation and is a poor antibiotic
Catalog No. 50-289-0219 Supplier Cayman Chemical Supplier No. 10009542500 MG
A powerful effector in both the TetR and revTetR transcriptional regulator systems, binding the Tet repressor 35-fold more strongly than Tet; does not act a general inhibitor of translation and is a poor antibiotic