Learn More
Medchemexpress LLC Anhydrotetracycline hydrochloride | 13803-65-1 | ≥98.0% | 462.88 | C22H23ClN2O7 | 10 MG
SDP
Catalog No. 5000228436
Encompass_Preferred
Anhydrotetracycline hydrochloride is a tetracycline-family antibiotic supplied as an analytical reference standard for laboratory research. It is commonly used as an effector in tetracycline-regulated gene expression systems and for analytical method development. The material is provided as the hydrochloride salt with molecular weight 462.88 g·mol⁻1 and chemical formula C22H23ClN2O7.
  • Reference standard for research and analytical applications.
  • Acts as an effector in tetracycline-regulated gene expression systems.
  • Supplied as a hydrochloride salt; formula C22H23ClN2O7 and MW 462.88 g·mol⁻1.
  • High reported purity (≥98%) suitable for analytical work.
  • Available in small laboratory pack sizes (e.g., 10 MG, 50 MG).
Catalog No. 50-002-28436 Supplier Medchemexpress LLC Supplier No. HY11866010MG
Anhydrotetracycline hydrochloride is a tetracycline-family antibiotic supplied as an analytical reference standard for laboratory research. It is commonly used as an effector in tetracycline-regulated gene expression systems and for analytical method development. The material is provided as the hydrochloride salt with molecular weight 462.88 g·mol⁻1 and chemical formula C22H23ClN2O7.
  • Reference standard for research and analytical applications.
  • Acts as an effector in tetracycline-regulated gene expression systems.
  • Supplied as a hydrochloride salt; formula C22H23ClN2O7 and MW 462.88 g·mol⁻1.
  • High reported purity (≥98%) suitable for analytical work.
  • Available in small laboratory pack sizes (e.g., 10 MG, 50 MG).