Learn More
Medchemexpress LLC Anhydrotetracycline hydrochloride | 13803-65-1 | 99.4% | 462.88 | 5 MG
SDP
Catalog No. 5000364634
Encompass_Preferred
Anhydrotetracycline hydrochloride is a tetracycline biosynthetic precursor and a potent competitive broad-spectrum inhibitor of tetracycline destructase enzymes. It also acts as an effector for tetracycline controlled gene expression systems in eukaryotic cells.
  • Inhibits Tet(50), Tet(X), and Tet(X)_3 degradations of tetracyclines
  • Inhibits Tet(50), Tet(X), and Tet(X)_3 degradations of chlortetracycline
  • Inhibits Tet(50), Tet(X), and Tet(X)_3 degradations of demeclocycline
  • Used in infection and bacterial infection research
Catalog No. 50-003-64634 Supplier Medchemexpress LLC Supplier No. 5MGHY118660
Anhydrotetracycline hydrochloride is a tetracycline biosynthetic precursor and a potent competitive broad-spectrum inhibitor of tetracycline destructase enzymes. It also acts as an effector for tetracycline controlled gene expression systems in eukaryotic cells.
  • Inhibits Tet(50), Tet(X), and Tet(X)_3 degradations of tetracyclines
  • Inhibits Tet(50), Tet(X), and Tet(X)_3 degradations of chlortetracycline
  • Inhibits Tet(50), Tet(X), and Tet(X)_3 degradations of demeclocycline
  • Used in infection and bacterial infection research