Learn More
Vector Laboratories Anhydrous DMF, 5 x 1.5 mL
SDP
Catalog No. 502045755
Encompass
Anhydrous DMF (Dimethylformamide) is used for solubilizing S-HyNic, S-4FB, and other SoluLINK linkers. Anhydrous DMF is packaged with molecular sieves to minimize water contamination and preserve quality.
Catalog No. 50-204-5755 Supplier Vector Laboratories Supplier No. S4001005
Anhydrous DMF (Dimethylformamide) is used for solubilizing S-HyNic, S-4FB, and other SoluLINK linkers. Anhydrous DMF is packaged with molecular sieves to minimize water contamination and preserve quality.