Learn More
Medchemexpress LLC Anhydrous sodium dihydrogen phosphate, for HPLC | 7558-80-7 | 99.9% | 10 G
SDP
Catalog No. 5000411011
Encompass_Preferred
Anhydrous sodium dihydrogen phosphate, for HPLC is widely used in molecular biology, biochemistry and chromatography. It can serve as a pH buffer and can be used as an excipient, such as a buffer or chelating agent, in pharmaceutical processes. Pharmaceutical excipients generally refer to inactive ingredients in pharmaceutical preparations, which can improve stability, solubility, and processability, and also affect the absorption, distribution, metabolism, and elimination processes of co-administered drugs.
  • Widely used in molecular biology, biochemistry, and chromatography
  • Can serve as a pH buffer
  • Used as an excipient in pharmaceutical preparations
  • Improves stability, solubility, and processability of pharmaceuticals
  • Affects ADME processes of co-administered drugs
Catalog No. 50-004-11011 Supplier Medchemexpress LLC Supplier No. HYB2243A10G
Anhydrous sodium dihydrogen phosphate, for HPLC is widely used in molecular biology, biochemistry and chromatography. It can serve as a pH buffer and can be used as an excipient, such as a buffer or chelating agent, in pharmaceutical processes. Pharmaceutical excipients generally refer to inactive ingredients in pharmaceutical preparations, which can improve stability, solubility, and processability, and also affect the absorption, distribution, metabolism, and elimination processes of co-administered drugs.
  • Widely used in molecular biology, biochemistry, and chromatography
  • Can serve as a pH buffer
  • Used as an excipient in pharmaceutical preparations
  • Improves stability, solubility, and processability of pharmaceuticals
  • Affects ADME processes of co-administered drugs