Learn More
Medchemexpress LLC Anhydrous sodium dihydrogen phosphate, for molecular biology | 7558-80-7 | 99.9% | 100 G
SDP
Catalog No. 5000409721
Encompass_Preferred
Anhydrous sodium dihydrogen phosphate, for molecular biology is an inorganic salt compound that can be used as a buffer and nutritional supplement in molecular biology experiments. It is also used in industry and laboratories as a buffer, nutritional supplement, cleaning agent, and in metal processing, pharmaceutical, chemical industries, water treatment, and environmental protection (as a purifying agent or precipitating agent for solutions).
  • Used as a buffer
  • Functions as a nutritional supplement
  • Employed as a cleaning agent
  • Utilized in metal processing
  • Applicable in pharmaceutical and chemical industries
  • Used in water treatment and environmental protection as a purifying or precipitating agent
Catalog No. 50-004-09721 Supplier Medchemexpress LLC Supplier No. HYB2243B100G
Anhydrous sodium dihydrogen phosphate, for molecular biology is an inorganic salt compound that can be used as a buffer and nutritional supplement in molecular biology experiments. It is also used in industry and laboratories as a buffer, nutritional supplement, cleaning agent, and in metal processing, pharmaceutical, chemical industries, water treatment, and environmental protection (as a purifying agent or precipitating agent for solutions).
  • Used as a buffer
  • Functions as a nutritional supplement
  • Employed as a cleaning agent
  • Utilized in metal processing
  • Applicable in pharmaceutical and chemical industries
  • Used in water treatment and environmental protection as a purifying or precipitating agent