Learn More
Medchemexpress LLC Anhydrous sodium dihydrogen phosphate, for molecular biology | 7558-80-7 | 99.9% | 50 G
SDP
Catalog No. 5000411012
Encompass_Preferred
Anhydrous sodium dihydrogen phosphate, for molecular biology is an inorganic salt compound that can be used as a buffer and nutritional supplement. It can also be used in molecular biology experiments. In a broader context, Anhydrous sodium dihydrogen phosphate is used in industry and laboratories as a buffer, nutritional supplement, and cleaning agent. It plays a role in certain metal processing, pharmaceutical, and chemical industries, and can be applied in water treatment and environmental protection as a purifying or precipitating agent.
  • Inorganic salt compound.
  • Can be used as a buffer.
  • Can be used as a nutritional supplement.
  • Can be used in molecular biology experiments.
  • Used as a cleaning agent.
  • Plays a role in metal processing, pharmaceutical, and chemical industries.
  • Can be used in water treatment and environmental protection as a purifying or precipitating agent.
Catalog No. 50-004-11012 Supplier Medchemexpress LLC Supplier No. HYB2243B50G
Anhydrous sodium dihydrogen phosphate, for molecular biology is an inorganic salt compound that can be used as a buffer and nutritional supplement. It can also be used in molecular biology experiments. In a broader context, Anhydrous sodium dihydrogen phosphate is used in industry and laboratories as a buffer, nutritional supplement, and cleaning agent. It plays a role in certain metal processing, pharmaceutical, and chemical industries, and can be applied in water treatment and environmental protection as a purifying or precipitating agent.
  • Inorganic salt compound.
  • Can be used as a buffer.
  • Can be used as a nutritional supplement.
  • Can be used in molecular biology experiments.
  • Used as a cleaning agent.
  • Plays a role in metal processing, pharmaceutical, and chemical industries.
  • Can be used in water treatment and environmental protection as a purifying or precipitating agent.