Learn More
Medchemexpress LLC HY-117102 10mg Medchemexpress, ANI-7 CAS:931417-26-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11710210MG
Encompass_Preferred
Medchemexpress, HY-117102 10mg ANI-7 CAS:931417-26-4 ANI-7 is an activator of aryl hydrocarbon receptor (AhR) pathway. ANI-7 inhibits the growth of multiple cancer cells, and potently and selectively inhibits the growth of MCF-7 breast cancer cells with a GI50 of 0.56 μM. ANI-7 induces CYP1-metabolizing mono-oxygenases by activating AhR pathway, and also induces DNA damage, checkpoint Kinase 2 (Chk2) activation, S-phase cell cycle arrest, and cell death in sensitive breast cancer cell lines[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9452