Learn More
Apexbio Technology LLC Anidulafungin(Synonyms: Eraxis, Ecalta, LY303366, VER-002, Anidulafungin acetate, V-echinocandin, Vicuron echinocandin), 10mM (in 1mL DMSO), CAS: 166663-25-8.
SDP
Catalog No. 5000060578
Encompass_Preferred
Anidulafungin (CAS 166663-25-8) also known as LY-303366 is a semisynthetic derivative of echinocandin B exhibiting antifungal activity It acts by inhibiting fungal cell wall synthesis through interference with the enzyme -(1 3)-D-glucan synthase In vitro analyses demonstrate that Anidulafungin inhibits various Candida species including C albicans C glabrata C tropicalis and C parapsilosis (MIC90 0 08 0 32 0 32 5 12 g/ml respectively) as well as Aspergillus species (MIC90 0 02 g/ml) Lack of significant efficacy was shown against Cryptococcus neoformans and Blastomyces dermatitidis This compound is useful for research exploring antifungal resistance mechanisms and therapeutics
Catalog No. 50-000-60578 Supplier Apexbio Technology LLC Supplier No. B12245.1
Anidulafungin (CAS 166663-25-8) also known as LY-303366 is a semisynthetic derivative of echinocandin B exhibiting antifungal activity It acts by inhibiting fungal cell wall synthesis through interference with the enzyme -(1 3)-D-glucan synthase In vitro analyses demonstrate that Anidulafungin inhibits various Candida species including C albicans C glabrata C tropicalis and C parapsilosis (MIC90 0 08 0 32 0 32 5 12 g/ml respectively) as well as Aspergillus species (MIC90 0 02 g/ml) Lack of significant efficacy was shown against Cryptococcus neoformans and Blastomyces dermatitidis This compound is useful for research exploring antifungal resistance mechanisms and therapeutics