Learn More
Abcam Anidulafungin, 5MG
Catalog No. 502671050
Encompass_Preferred
MW 1140.2 g/mol, Purity >=95%. Inhibits fungal (1→3)-β-D-glucan synthase. Potent in vitro activity against Aspergillus and Candida species (MIC₅₀ of 0.03 to 4 μg/ml), including those resistant to fluconazole or amphotericin B. Effective in treating esophageal candidiasis in clinical trials.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1050 Supplier Abcam Supplier No. AB2898515MG
MW 1140.2 g/mol, Purity >=95%. Inhibits fungal (1→3)-β-D-glucan synthase. Potent in vitro activity against Aspergillus and Candida species (MIC₅₀ of 0.03 to 4 μg/ml), including those resistant to fluconazole or amphotericin B. Effective in treating esophageal candidiasis in clinical trials.
The product is subject to the following: Abcam Restricted Use Statement