Learn More
Selleck Chemical LLC Anidulafungin (LY303366) 10mg 166663-25-8
SDP
Catalog No. 501363853
Encompass_Preferred
Anidulafungin (LY303366), an echinocandin derivative, inhibits glucan synthase activity, used as an antifungal drug. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3853 Supplier Selleck Chemical LLC Supplier No. S428610MG
Anidulafungin (LY303366), an echinocandin derivative, inhibits glucan synthase activity, used as an antifungal drug. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.