Learn More
Medchemexpress LLC Anifrolumab, 1mg | CAS No. : 1326232-46-5
SDP
Supplier:  Medchemexpress LLC HYP991681MG
Encompass_Preferred
Anifrolumab is a type I interferon (IFN) receptor antagonist, a human monoclonal antibody. Anifrolumab blocks the activity of type I interferon. Anifrolumab can be used in systemic lupus erythematosus (SLE) research.
Catalog No. NC2890446