Learn More
Biotium Aniline, 10X in acetate buffer
Catalog No. 501964834
Encompass_Preferred
Aniline, 10X in acetate buffer is designed to use as catalytic reagent for the optimal coupling of protein carbonyls with aminooxy compounds.
Catalog No. 50-196-4834 Supplier Biotium Supplier No. 91057
Add to Cart
Add to Cart
Aniline, 10X in acetate buffer is designed to use as catalytic reagent for the optimal coupling of protein carbonyls with aminooxy compounds.