Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aniline-2,3,4,5,6-d5 99+ atom% D, Thermo Scientific™

Catalog No. AC174700010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC174700010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC174700010 Supplier Thermo Scientific Chemicals Supplier No. 174700010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4165-61-1
Molecular Formula C6H7N
Molecular Weight (g/mol) 98.16
MDL Number MFCD00044420
InChI Key PAYRUJLWNCNPSJ-RALIUCGRSA-N
Synonym benzen-d5-amine, aniline-d5, aniline d5, aniline-2,3,4,5,6-d5, 2 h? aniline, phenyl-d5-amine, aniline #, 2,3,4,5,6-2h5 aniline, 2h5 aniline, aniline-2'3'4'5'6-d5
PubChem CID 123211
IUPAC Name (²H₅)aniline
SMILES [2H]C1=C([2H])C([2H])=C(N)C([2H])=C1[2H]

Specifications

Physical Form Liquid
Color Brown-Yellow to Orange
Quantity 1 g
Deuterium (D) 99.0% min.
Linear Formula C6D5NH2
Formula Weight 98.17
Chemical Name or Material Aniline-2,3,4,5,6-d5, 99+ atom % D
Hazard Category HYGROSCOPIC
Hazard Statement LIGHT SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.