Learn More
Aniline, 99.5%, extra pure
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -6.2°C |
| Color | Brown to Colorless |
| Density | 1.0213g/mL |
| Boiling Point | 181.0°C to 185.0°C |
| Flash Point | 76°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99.4% min. (GC) |
| Packaging | Plastic drum |
| Refractive Index | 1.5845 to 1.5865 |
| Linear Formula | C6H5NH2 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Suspected of causing cancer. Causes damage to organs through prolonged or repeated exposure. Suspected of causing genetic defects. Toxic in contact with skin. Toxic if swallowed. Causes serious eye dam |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF |
| EINECSNumber | 200-539-3 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.