Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Aniline Blue, C.I. DcK-11 25 GR
SDP

Catalog No. 50980184
Encompass_Preferred

(Acid Blue 22; China Blue; Cotton Blue; Marine Blue V; Soluble Blue 3M, 2R; Water Blue)
CAS #28631-66-5 C₃₂H₂₅N₃O₉S₃Na₂ F.W. 737.74
Solubility: 50% Water; 0.0% Alcohol; 4.5% Cellosolve; 7.8% Glycol; 0.0% Xylene

Aniline Blue Disodium Salt is used in connective tissue staining procedures to stain collagen. Used in differential staining (Snodgress el al., Lab Invest. 26:329, 1972).

Catalog No. 50-980-184 Supplier Electron Microscopy Sciences Supplier No. 10840
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Acid Blue 22; China Blue; Cotton Blue; Marine Blue V; Soluble Blue 3M, 2R; Water Blue)
CAS #28631-66-5 C₃₂H₂₅N₃O₉S₃Na₂ F.W. 737.74
Solubility: 50% Water; 0.0% Alcohol; 4.5% Cellosolve; 7.8% Glycol; 0.0% Xylene

Aniline Blue Disodium Salt is used in connective tissue staining procedures to stain collagen. Used in differential staining (Snodgress el al., Lab Invest. 26:329, 1972).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.