Learn More
Medchemexpress LLC Aniline blue sodium | 28631-66-5 | MFCD00036130 | 737.73 g·mol⁻¹ | C32H25N3Na2O9S3 | 25 G
SDP
Catalog No. 5000283506
Encompass_Preferred
Aniline blue sodium is a water-soluble dye used as a biochemical reagent and biological stain to visualize nucleic acids and proteins in laboratory applications such as electrophoresis and microscopy. It is supplied as a purple-to-black solid for research use and is available in multiple pack sizes.
  • Water-soluble dye suitable for aqueous staining protocols.
  • Effective for visualization of nucleic acids and proteins.
  • Compatible with electrophoresis and microscopic staining techniques.
  • Available in multiple pack sizes for laboratory needs.
  • Supplied as a stable solid with defined molecular properties.
Catalog No. 50-002-83506 Supplier Medchemexpress LLC Supplier No. HYW13391925G
Aniline blue sodium is a water-soluble dye used as a biochemical reagent and biological stain to visualize nucleic acids and proteins in laboratory applications such as electrophoresis and microscopy. It is supplied as a purple-to-black solid for research use and is available in multiple pack sizes.
  • Water-soluble dye suitable for aqueous staining protocols.
  • Effective for visualization of nucleic acids and proteins.
  • Compatible with electrophoresis and microscopic staining techniques.
  • Available in multiple pack sizes for laboratory needs.
  • Supplied as a stable solid with defined molecular properties.