Learn More
Medchemexpress LLC Aniline blue, sodium salt | 28631-66-5 | MFCD00036130 | 737.73 g·mol⁻¹ | C32H25N3Na2O9S3 | 10 G
SDP
Catalog No. 5000283369
Encompass_Preferred
Aniline blue, sodium salt is a water-soluble biological dye supplied as a powder for research use. It is used in histological and biochemical staining protocols to detect nucleic acids and proteins, producing strong coloration for microscopy and colorimetric assays.
  • Water-soluble dye suitable for staining nucleic acids and proteins.
  • Powdered form for easy preparation of aqueous solutions.
  • Compatible with common histological and biochemical staining protocols.
  • Produces strong staining intensity for clear visualization.
  • Supplied for laboratory research applications.
Catalog No. 50-002-83369 Supplier Medchemexpress LLC Supplier No. HYW13391910G
Aniline blue, sodium salt is a water-soluble biological dye supplied as a powder for research use. It is used in histological and biochemical staining protocols to detect nucleic acids and proteins, producing strong coloration for microscopy and colorimetric assays.
  • Water-soluble dye suitable for staining nucleic acids and proteins.
  • Powdered form for easy preparation of aqueous solutions.
  • Compatible with common histological and biochemical staining protocols.
  • Produces strong staining intensity for clear visualization.
  • Supplied for laboratory research applications.