Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aniline (Certified ACS), Fisher Chemical™

Catalog No. A740I500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A740I-500 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A740I-500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product darkens to a reddish-brown color on storage

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-53-3
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.13
MDL Number MFCD00007629
InChI Key PAYRUJLWNCNPSJ-UHFFFAOYSA-N
Synonym benzenamine, phenylamine, aminobenzene, aminophen, anilin, kyanol, arylamine, cyanol, benzeneamine, benzidam
PubChem CID 6115
ChEBI CHEBI:17296
IUPAC Name aniline
SMILES NC1=CC=CC=C1

Specifications

Melting Point -6.2°C
Color Yellow
Density 1.021g/cm³
pH 8.8
Boiling Point 181°C
Flash Point 76°C
Ignition Residue 0.005% max.
Packaging Glass Bottle
Linear Formula C6H5NH2
Quantity 500 mL
UN Number 1547
Vapor Pressure 0.5mmHg at 20°C
Formula Weight 93.13
Percent Purity ≥99.0%
Grade Certified ACS
Physical Form Liquid
Chemical Name or Material Aniline
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Combustible liquid. Toxic by inhalation, in contact with skin and if swallowed. Risk of serious damage to eyes. May cause an allergic skin reaction. May cause skin and respiratory tract irritation. Possible risks of irreversible effects. Danger of serious damage to health by prolonged exposure. Very toxic to aquatic organisms. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:2
Flammability:2
Instability:0
DOTInformation DOT Class 6.1, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.