Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Aniline hydrochloride

Catalog No. 501804610
Encompass
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-180-4610 50 g
50-180-4611 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-180-4610 Supplier Sigma Aldrich Supplier No. A852450G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Aniline hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

CAS 142-04-1
Melting Point 196°C to 198°C (lit.)
Molecular Formula C6H7N -+ HCl
Linear Formula C6H5NH2 -+ HCl
Quantity 50 g
MDL Number MFCD00012958
Molecular Weight (g/mol) 129.59
Percent Purity ≥99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.