Learn More
Medchemexpress LLC Aniline-PEG3-C1-Boc | 2115897-27-1 | 355.43 | C18H29NO6 | 10 MG
SDP
Catalog No. 5000203030
Encompass_Preferred
Aniline-PEG3-C1-Boc is a chemical intermediate used in the synthesis of cytotoxic drug-linker polymers and antibody-drug conjugates (ADCs). It is supplied as a liquid and is intended for research and development applications in ADC linker chemistry.
  • Intermediate for cytotoxic drug-linker polymer synthesis.
  • Used in antibody-drug conjugate (ADC) construction.
  • Molecular formula C18H29NO6 and molecular weight 355.43 g/mol.
  • Available in small milligram pack sizes suitable for research (5 mg-500 mg).
  • Store pure material at -20°C; follow solvent-specific storage instructions.
Catalog No. 50-002-03030 Supplier Medchemexpress LLC Supplier No. HY16463710MG
Aniline-PEG3-C1-Boc is a chemical intermediate used in the synthesis of cytotoxic drug-linker polymers and antibody-drug conjugates (ADCs). It is supplied as a liquid and is intended for research and development applications in ADC linker chemistry.
  • Intermediate for cytotoxic drug-linker polymer synthesis.
  • Used in antibody-drug conjugate (ADC) construction.
  • Molecular formula C18H29NO6 and molecular weight 355.43 g/mol.
  • Available in small milligram pack sizes suitable for research (5 mg-500 mg).
  • Store pure material at -20°C; follow solvent-specific storage instructions.