Learn More
Medchemexpress LLC Aniline-PEG3-C1-Boc | 2115897-27-1 | 355.43 g/mol | C18H29NO6 | 100 MG
SDP
Catalog No. 5000202532
Encompass_Preferred
Aniline-PEG3-C1-Boc (compound D-1) is a Boc-protected aniline PEG3 linker intermediate used in the synthesis of cytotoxic drug linker polymers and antibody-drug conjugates (ADCs).
  • Boc-protected aniline PEG3 linker for synthetic chemistry
  • Applicable to antibody-drug conjugate (ADC) linker construction
  • Supplied in small laboratory quantities for research use
  • Available in multiple package sizes, including 100 mg
  • Contains CAS number 2115897-27-1 and molecular weight 355.43 g/mol
Catalog No. 50-002-02532 Supplier Medchemexpress LLC Supplier No. HY164637100MG
Aniline-PEG3-C1-Boc (compound D-1) is a Boc-protected aniline PEG3 linker intermediate used in the synthesis of cytotoxic drug linker polymers and antibody-drug conjugates (ADCs).
  • Boc-protected aniline PEG3 linker for synthetic chemistry
  • Applicable to antibody-drug conjugate (ADC) linker construction
  • Supplied in small laboratory quantities for research use
  • Available in multiple package sizes, including 100 mg
  • Contains CAS number 2115897-27-1 and molecular weight 355.43 g/mol