Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aniline-PEG3-C1-Boc | 2115897-27-1 | 355.43 g/mol | C18H29NO6 | 25 MG
SDP

Supplier:  Medchemexpress LLC HY16463725MG

Encompass_Preferred

Aniline-PEG3-C1-Boc is a protected aniline-PEG3 intermediate used as a building block in cytotoxic drug-linker polymer and antibody-drug conjugate (ADC) synthesis.

  • Intermediate for cytotoxic drug-linker polymer synthesis.
  • Applicable for antibody-drug conjugate (ADC) synthesis.
  • Molecular formula: C18H29NO6.
  • Molecular weight: 355.43 g/mol.
  • Provided in small-scale research quantities (e.g., 25 mg).

Catalog No. 50-002-01932


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.