Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Aniline-PEG3-C1-Boc | 2115897-27-1 | 355.43 g/mol | C18H29NO6 | 50 MG
SDP

Catalog No. 5000203048
Encompass_Preferred

Aniline-PEG3-C1-Boc is a Boc-protected aniline PEG3 intermediate used as a building block in linker-drug polymers and in the synthesis of antibody-drug conjugates (ADCs). It is supplied for research use and is highly soluble in DMSO.

  • CAS number: 2115897-27-1
  • Molecular formula: C18H29NO6
  • Molecular weight: 355.43 g/mol
  • Solubility: 350 mg/mL in DMSO (ultrasonication assisted)
  • Storage: pure form -20°C (stable for 3 years); in solvent -80°C for 6 months
  • Typical application: intermediate for ADC linker and conjugate synthesis

Catalog No. 50-002-03048 Supplier Medchemexpress LLC Supplier No. HY16463750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aniline-PEG3-C1-Boc is a Boc-protected aniline PEG3 intermediate used as a building block in linker-drug polymers and in the synthesis of antibody-drug conjugates (ADCs). It is supplied for research use and is highly soluble in DMSO.

  • CAS number: 2115897-27-1
  • Molecular formula: C18H29NO6
  • Molecular weight: 355.43 g/mol
  • Solubility: 350 mg/mL in DMSO (ultrasonication assisted)
  • Storage: pure form -20°C (stable for 3 years); in solvent -80°C for 6 months
  • Typical application: intermediate for ADC linker and conjugate synthesis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.