Learn More
Sigma Aldrich Fine Chemicals Biosciences Aniline ReagentPlusR 99
Catalog No. 502891513
Encompass
Aniline is an aromatic amine. It is a key industrial intermediate, widely employed for the preparation of dyes, rubber, resins and polymers. Complete vibrational assignements of aniline and its deuterated forms (aniline-NHD and aniline-ND2) have been reported.1 p-aminodiphenylamine (ADPA) has been reported to be formed as a major intermediate during the electropolymerizatrion of aniline. Its viscosity has been reported to be 3.1457cp at 30C. Quantitative transformation of nitrobenzene to aniline has been carried out by employing functionalized plasmonic Au/TiO2 photocatalyst along with a Ag co-catalyst. Aniline can be prepared from nitrobenzene, via hydrogenation in the presence of Cu, Ni, Pt, Pd and Au (catalyst). It can also be prepared by photocatalytic reduction of nitrobenzene in the presence of WO3-Ag hybrid nanowires.
Catalog No. 50-289-1513 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 13293418L
Aniline is an aromatic amine. It is a key industrial intermediate, widely employed for the preparation of dyes, rubber, resins and polymers. Complete vibrational assignements of aniline and its deuterated forms (aniline-NHD and aniline-ND2) have been reported.1 p-aminodiphenylamine (ADPA) has been reported to be formed as a major intermediate during the electropolymerizatrion of aniline. Its viscosity has been reported to be 3.1457cp at 30C. Quantitative transformation of nitrobenzene to aniline has been carried out by employing functionalized plasmonic Au/TiO2 photocatalyst along with a Ag co-catalyst. Aniline can be prepared from nitrobenzene, via hydrogenation in the presence of Cu, Ni, Pt, Pd and Au (catalyst). It can also be prepared by photocatalytic reduction of nitrobenzene in the presence of WO3-Ag hybrid nanowires.