Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anilinoacetaldehyde diethyl acetal, 98%, Thermo Scientific™

Catalog No. AAA1158714
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1158714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1158714 Supplier Thermo Scientific Chemicals Supplier No. A1158714

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22758-34-5
Molecular Formula C12H19NO2
Molecular Weight (g/mol) 209.289
MDL Number MFCD00051601
InChI Key DHGUGEBXVGPRRD-UHFFFAOYSA-N
Synonym anilinoacetaldehyde diethyl acetal, n-2,2-diethoxyethyl aniline, n-2,2-diethoxyethyl benzenamine, n-phenylaminoacetaldehyde diethyl acetal, acmc-1ch3s, 2,2-diethoxyethyl phenylamine, n-2,2-diethoxyethyl aniline #, benzenamine,n-2,2-diethoxyethyl, n-phenyl-2-aminoacetaldehyde diethylacetal
PubChem CID 519978
IUPAC Name N-(2,2-diethoxyethyl)aniline
SMILES CCOC(CNC1=CC=CC=C1)OCC

Specifications

Boiling Point 153°C to 154°C (12 mmHg)
Quantity 25 g
Beilstein 2806325
Formula Weight 209.29
Percent Purity 98%
Chemical Name or Material Anilinoacetaldehyde diethyl acetal
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.