Learn More
STEMCELL Technologies CryoStor CSB, Size: 100 mL
SDP
Catalog No. 502045486
Encompass
Reduce the level of cryopreservation-induced delayed-onset cell death and maximize post-thaw cell recovery, viability, and function following cryopreservation, with CryoStor CSB, an intracellular-like base formulation from the CryoStor preservation media series. Serum-free, animal component-free, and cGMP-manufactured, this chemically defined medium is sterile and tested for endotoxins and cell-based release. CryoStor CSB is formulated with USP-grade components and does not contain DMSO. Visit STEMCELL.com for more information.
Catalog No. 50-204-5486 Supplier STEMCELL Technologies Supplier No. 1000237
Reduce the level of cryopreservation-induced delayed-onset cell death and maximize post-thaw cell recovery, viability, and function following cryopreservation, with CryoStor CSB, an intracellular-like base formulation from the CryoStor preservation media series. Serum-free, animal component-free, and cGMP-manufactured, this chemically defined medium is sterile and tested for endotoxins and cell-based release. CryoStor CSB is formulated with USP-grade components and does not contain DMSO. Visit STEMCELL.com for more information.