Learn More
STEMCELL Technologies StemSpan™-ACF Erythroid Expansion Medium, Size: 500 mL Bottle
SDP
Catalog No. 502045468
Encompass
StemSpan™-ACF Erythroid Expansion Medium (ACF-E) is an animal component-free (ACF) medium for the in vitro culture and expansion of human hematopoietic cells. When supplemented with StemSpan™ Erythroid Expansion Supplement (100X; Catalog #02692), this medium supports the expansion and erythroid differentiation of human hematopoietic progenitor cells into CD71+ and Glycophorin A+ erythroblasts in ACF culture conditions. This medium only contains recombinant proteins and synthetic components; it does not contain serum or other human- or animal-derived components.
Catalog No. 50-204-5468 Supplier STEMCELL Technologies Supplier No. 09860
StemSpan™-ACF Erythroid Expansion Medium (ACF-E) is an animal component-free (ACF) medium for the in vitro culture and expansion of human hematopoietic cells. When supplemented with StemSpan™ Erythroid Expansion Supplement (100X; Catalog #02692), this medium supports the expansion and erythroid differentiation of human hematopoietic progenitor cells into CD71+ and Glycophorin A+ erythroblasts in ACF culture conditions. This medium only contains recombinant proteins and synthetic components; it does not contain serum or other human- or animal-derived components.