Learn More
Sigma Aldrich Fine Chemicals Biosciences Animal feeding needle ster
Supplier:  Sigma Aldrich Fine Chemicals Biosciences CAD9924B100EA
Encompass
flexible PTfE tube covered at the hub end with type 304 stainless steel preventing tube damage due to biting during usePTfE tube is softer than other tubing types very flexible and translucent for easy confirmation of reagent deliveryPlastic Luer hubs and soft silicone tipsNon-toxic and no-pyrogenicPackaged 5 needles per sterile Tyvek peel pouch - 20 pouches per box
Catalog No. 50-290-5779