Learn More
Sigma Aldrich Fine Chemicals Biosciences Animal feeding needle ster
Supplier:  Sigma Aldrich Fine Chemicals Biosciences CAD9931100EA
Encompass
PTFE tube is softer than other tubing types very flexible and translucent for easy confirmation of reagent deliveryPlastic Luer hubs and soft silicone tipsNon-toxic and no-pyrogenicPackaged 5 needles per sterile Tyvek peel pouch - 20 pouches per box
Catalog No. 50-290-8747