Learn More
Sigma Aldrich Fine Chemicals Biosciences Animal feeding needle ster
Catalog No. 502905523
Encompass
Designed to enable the researcher to shape the needle to specific needsNeedles come straight and are easily bent to any desired shapeMade of type 304 stainless steel tubingPlastic Luer hubsSilicone tips for smoothest possible insertionPackaged 5 needles per sterile Tyvek peel pouch - 20 pouches per box
Catalog No. 50-290-5523 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CAD9921100EA
Designed to enable the researcher to shape the needle to specific needsNeedles come straight and are easily bent to any desired shapeMade of type 304 stainless steel tubingPlastic Luer hubsSilicone tips for smoothest possible insertionPackaged 5 needles per sterile Tyvek peel pouch - 20 pouches per box