Learn More
Medchemexpress LLC Nerve growth factor-β (beta-NGF) | 95.0% | 10UG
SDP
Catalog No. 5000177452
Encompass_Preferred
Recombinant, animal-free human beta-nerve growth factor (beta-NGF) expressed with a C-terminal His tag and produced in E. coli. The preparation corresponds to amino acids S122-A241, is provided at small research scale, and is characterized by SDS-PAGE analysis for molecular weight and purity.
  • Animal-free recombinant human protein suitable for research
  • Corresponds to amino acids S122-A241 of human beta-NGF
  • C-terminal His tag for detection and purification
  • Purity 95.0% as analyzed by SDS-PAGE
  • Approximate molecular weight 11.0 kDa (SDS-PAGE)
  • Supplied in small research quantity (10 μg)
  • For cell culture use only
Catalog No. 50-001-77452 Supplier Medchemexpress LLC Supplier No. HYP700018AF10UG
Recombinant, animal-free human beta-nerve growth factor (beta-NGF) expressed with a C-terminal His tag and produced in E. coli. The preparation corresponds to amino acids S122-A241, is provided at small research scale, and is characterized by SDS-PAGE analysis for molecular weight and purity.
  • Animal-free recombinant human protein suitable for research
  • Corresponds to amino acids S122-A241 of human beta-NGF
  • C-terminal His tag for detection and purification
  • Purity 95.0% as analyzed by SDS-PAGE
  • Approximate molecular weight 11.0 kDa (SDS-PAGE)
  • Supplied in small research quantity (10 μg)
  • For cell culture use only