Learn More
Cell Signaling Technology Animal-Free Blocking Solution, 5X, 250 mL
SDP
Catalog No. 502050431
Encompass
Animal-Free Blocking Solution (5X) is a plant-based reagent developed to minimize background in chromogenic immunohistochemical assays and western blotting. This formulation is designed to replace traditional blocking agents such as normal goat serum, BSA, or non-fat dry milk, providing a solution free of animal-derived proteins.
  • Contains no animal-derived proteins
  • Formulated for use in western blotting and immunohistochemistry
  • Concentrated 5X formulation requiring dilution to 1X with deionized water prior to use
  • Stable for 36 months when stored at +4°C and protected from light
Catalog No. 50-205-0431 Supplier Cell Signaling Technology Supplier No. 15019L
Animal-Free Blocking Solution (5X) is a plant-based reagent developed to minimize background in chromogenic immunohistochemical assays and western blotting. This formulation is designed to replace traditional blocking agents such as normal goat serum, BSA, or non-fat dry milk, providing a solution free of animal-derived proteins.
  • Contains no animal-derived proteins
  • Formulated for use in western blotting and immunohistochemistry
  • Concentrated 5X formulation requiring dilution to 1X with deionized water prior to use
  • Stable for 36 months when stored at +4°C and protected from light