Learn More
Medchemexpress LLC Bone morphogenetic protein 15, human recombinant (C-terminal 6xHis tag) | ≥98.0% | 10UG
SDP
Supplier:  Medchemexpress LLC HYP700024AF10UG
Encompass_Preferred
Recombinant human bone morphogenetic protein 15 (BMP-15) is provided as a lyophilized, C-terminal His-tagged protein expressed in E. coli for research applications. The animal-free formulation is characterized for purity, low endotoxin, and measured biological activity, and is intended for use in cell culture and biochemical assays.
  • High purity (≥98%) as determined by reducing SDS-PAGE.
  • Low endotoxin level (<0.1 EU/μg) by LAL test.
  • Biologically active; ED50 = 16.1 ng/mL in ATDC5 alkaline phosphatase assay.
  • Recombinant expression in E. coli with C-terminal 6xHis tag.
  • Lyophilized formulation with trehalose for improved stability.
  • Stable at -20°C; reconstitute to 100-200 μg/mL and aliquot for long-term storage.
Catalog No. 50-001-77682