Learn More
Medchemexpress LLC Bone morphogenetic protein 15 (BMP-15) | >98.0% | 2UG
SDP
Catalog No. 5000177653
Encompass_Preferred
Animal-free recombinant human bone morphogenetic protein 15 (BMP-15) is a C-terminal His-tagged growth factor fragment (UniProt O95972, Q268-R392) produced in E. coli and supplied for research use. It is intended for cell culture and in vitro assays and is accompanied by a batch certificate of analysis demonstrating high purity.
  • High purity (batch COA ≥98.0%).
  • C-terminal His tag for detection and purification.
  • Produced in E. coli using animal-free processes.
  • Corresponds to human residues Q268-R392.
  • Observed size ≈13-18 kDa by SDS-PAGE.
  • For research use only; not intended for human use.
Catalog No. 50-001-77653 Supplier Medchemexpress LLC Supplier No. HYP700024AF2UG
Animal-free recombinant human bone morphogenetic protein 15 (BMP-15) is a C-terminal His-tagged growth factor fragment (UniProt O95972, Q268-R392) produced in E. coli and supplied for research use. It is intended for cell culture and in vitro assays and is accompanied by a batch certificate of analysis demonstrating high purity.
  • High purity (batch COA ≥98.0%).
  • C-terminal His tag for detection and purification.
  • Produced in E. coli using animal-free processes.
  • Corresponds to human residues Q268-R392.
  • Observed size ≈13-18 kDa by SDS-PAGE.
  • For research use only; not intended for human use.