Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Animal-free CCL4 protein, mouse (His) | >95.0% | 50UG
SDP

Catalog No. 5000178075
Encompass_Preferred

Animal-Free CCL4 Protein, Mouse (His) is a recombinant mouse chemokine produced in E. coli and supplied for research and cell culture use. The product is provided in multiple sizes (including 50 μg), carries an N-terminal His tag, and is characterized by high purity and defined sequence for use in signaling, chemotaxis, and receptor-binding studies.

  • Recombinant mouse CCL4 expressed in E. coli.
  • N-terminal 6xHis tag for detection and purification.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Available in multiple sizes to suit assay scale.
  • Store at -20°C; freeze aliquots for extended storage.

Catalog No. 50-001-78075 Supplier Medchemexpress LLC Supplier No. HYP700165AF50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Animal-Free CCL4 Protein, Mouse (His) is a recombinant mouse chemokine produced in E. coli and supplied for research and cell culture use. The product is provided in multiple sizes (including 50 μg), carries an N-terminal His tag, and is characterized by high purity and defined sequence for use in signaling, chemotaxis, and receptor-binding studies.

  • Recombinant mouse CCL4 expressed in E. coli.
  • N-terminal 6xHis tag for detection and purification.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Available in multiple sizes to suit assay scale.
  • Store at -20°C; freeze aliquots for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.