Learn More
Medchemexpress LLC Animal-Free CNTF Mo 2ug
SDP
Supplier:  Medchemexpress LLC HYP74242AF2UG
Encompass_Preferred
CNTF is a pluripotent neurotrophic factor belonging to the IL-6 cytokine family[1] CNTF could protect retinal cone and rod photoreceptors[2] CNTF has neuroprotective effects on a variety of central and also peripheral nervous system neurons[3] Animal-Free CNTF Protein Mouse (His) is produced by E coli (M1-M198)with C-terminal His-tag This product is for cell culture use only
Catalog No. 50-001-78859