Learn More
AVANTOR MASTERFLEX (VWR LLC) Animal Free Female Luer Fittings, 3/16"", PP, 25/pk
Catalog No. NC1338718
Encompass
Animal Free Female Luer Fittings, 3/16", Polypropylene, 25/pk, Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization. (not recommended for autoclaving). Sold in packs of 25.
Catalog No. NC1338718 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3080004
Animal Free Female Luer Fittings, 3/16", Polypropylene, 25/pk, Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization. (not recommended for autoclaving). Sold in packs of 25.