Learn More
Medchemexpress LLC Animal-Free FGF-2 Protein, Mouse (His) | 96% | Approximately 19 kDa | 2 UG
SDP
Catalog No. 5000399986
Encompass_Preferred
FGF-2 is a member of the fibroblast family involved in bone healing, cartilage repair, bone repair, and nerve regeneration. It also acts as a mitotic promoter, accelerating cell proliferation. FGF-2 regulates immune processes by specifically targeting tyrosine kinase receptors and activating the FGF/FGFR signaling pathway. This product is for cell culture use only.
  • Consists of 144 amino acids
  • Produced by E. coli with tag-free protein
  • Offers high consistency and stability
  • Does not use or contact any animal-derived materials and reagents
Catalog No. 50-003-99986 Supplier Medchemexpress LLC Supplier No. HYP73052AF2UG
FGF-2 is a member of the fibroblast family involved in bone healing, cartilage repair, bone repair, and nerve regeneration. It also acts as a mitotic promoter, accelerating cell proliferation. FGF-2 regulates immune processes by specifically targeting tyrosine kinase receptors and activating the FGF/FGFR signaling pathway. This product is for cell culture use only.
  • Consists of 144 amino acids
  • Produced by E. coli with tag-free protein
  • Offers high consistency and stability
  • Does not use or contact any animal-derived materials and reagents