Learn More
Medchemexpress LLC Animal-free FGF-8a protein, human (His) | >95.0% | 50UG
SDP
Catalog No. 5000177560
Encompass_Preferred
Animal-free FGF-8a protein, human (His) is a recombinant human FGF-8a isoform produced in Escherichia coli and supplied as a lyophilized powder with a C-terminal His tag. It is intended for cell culture applications, supplied as a 50 μg vial, with sequence range P55075-2 (Q23-R204), ≥95.0% purity, and an approximate molecular weight of 23.0 kDa. Reconstitute to 100-200 μg/mL as recommended.
  • Recombinant human FGF-8a isoform with C-terminal His tag.
  • Produced in Escherichia coli and supplied as lyophilized powder.
  • High purity (≥95.0%) suitable for in vitro cell culture work.
  • Approximate molecular weight ~23.0 kDa by SDS-PAGE.
  • Recommended reconstitution concentration 100-200 μg/mL.
Catalog No. 50-001-77560 Supplier Medchemexpress LLC Supplier No. HYP700069AF50UG
Animal-free FGF-8a protein, human (His) is a recombinant human FGF-8a isoform produced in Escherichia coli and supplied as a lyophilized powder with a C-terminal His tag. It is intended for cell culture applications, supplied as a 50 μg vial, with sequence range P55075-2 (Q23-R204), ≥95.0% purity, and an approximate molecular weight of 23.0 kDa. Reconstitute to 100-200 μg/mL as recommended.
  • Recombinant human FGF-8a isoform with C-terminal His tag.
  • Produced in Escherichia coli and supplied as lyophilized powder.
  • High purity (≥95.0%) suitable for in vitro cell culture work.
  • Approximate molecular weight ~23.0 kDa by SDS-PAGE.
  • Recommended reconstitution concentration 100-200 μg/mL.