Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Animal-free G-CSF protein, human (His) | >95.0% | 2UG
SDP

Catalog No. 5000177524
Encompass_Preferred

Recombinant animal-free human granulocyte colony-stimulating factor (G-CSF) with an N-terminal 6xHis tag, expressed in E. coli and supplied lyophilized for cell-culture research applications.

  • Monomeric protein corresponding to amino acids T31-P204.
  • Predicted molecular mass 19.5 kDa; apparent ~21 kDa by SDS-PAGE.
  • Purity >95.0% as determined by reducing SDS-PAGE.
  • Endotoxin <0.1 EU per 1 μg (LAL method).
  • Lyophilized formulation in PBS, pH 7.4, with trehalose.
  • Store at -20°C; after reconstitution stable 4°C for 1 week, -20°C for longer (aliquot and freeze at -20°C or -80°C).
  • For cell culture use only.

Catalog No. 50-001-77524 Supplier Medchemexpress LLC Supplier No. HYP700083AF2UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant animal-free human granulocyte colony-stimulating factor (G-CSF) with an N-terminal 6xHis tag, expressed in E. coli and supplied lyophilized for cell-culture research applications.

  • Monomeric protein corresponding to amino acids T31-P204.
  • Predicted molecular mass 19.5 kDa; apparent ~21 kDa by SDS-PAGE.
  • Purity >95.0% as determined by reducing SDS-PAGE.
  • Endotoxin <0.1 EU per 1 μg (LAL method).
  • Lyophilized formulation in PBS, pH 7.4, with trehalose.
  • Store at -20°C; after reconstitution stable 4°C for 1 week, -20°C for longer (aliquot and freeze at -20°C or -80°C).
  • For cell culture use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.