Learn More
Medchemexpress LLC Animal-free GDNF protein, mouse (His) | ≥98.0% | 10UG
SDP
Supplier:  Medchemexpress LLC HYP700179AF10UG
Encompass_Preferred
Recombinant animal-free mouse GDNF (His-tag) supplied as a lyophilized powder for research use. Expressed in E. coli and characterized by SDS-PAGE and endotoxin testing, this protein is intended to support survival and maintenance of dopaminergic neurons and for use in cell culture and organoid applications.
  • Lyophilized recombinant mouse GDNF with C-terminal His tag.
  • Produced in E. coli; animal-free expression system.
  • Purity ≥98% by reducing SDS-PAGE.
  • Endotoxin <0.1 EU/μg (LAL).
  • Recommended reconstitution to 100-200 μg/mL; add carrier for long-term storage.
Catalog No. 50-001-77834