Learn More
Medchemexpress LLC Animal-free GDNF protein, mouse (His) | ≥98.0% | 50UG
SDP
Catalog No. 5000177746
Encompass_Preferred
Animal-Free GDNF Protein, Mouse (His) is a recombinant mouse glial cell line-derived neurotrophic factor fragment (residues S78-I211) produced in E. coli with a C-terminal 6xHis tag. Supplied as a highly purified protein (≥98.0%), it is intended for in vitro research and cell culture applications; store at -20°C and follow reconstitution and aliquoting guidance for stability.
  • Recombinant mouse GDNF fragment corresponding to residues S78-I211.
  • C-terminal 6xHis tag for detection and purification.
  • Produced in E. coli as an animal-free recombinant protein.
  • High purity (≥98.0%) for consistent experimental results.
  • Stable at -20°C; after reconstitution stable at 4°C for 1 week or -20°C for long-term (with carrier protein).
  • Available in small research quantities (2 μg, 10 μg, 50 μg).
Catalog No. 50-001-77746 Supplier Medchemexpress LLC Supplier No. HYP700179AF50UG
Animal-Free GDNF Protein, Mouse (His) is a recombinant mouse glial cell line-derived neurotrophic factor fragment (residues S78-I211) produced in E. coli with a C-terminal 6xHis tag. Supplied as a highly purified protein (≥98.0%), it is intended for in vitro research and cell culture applications; store at -20°C and follow reconstitution and aliquoting guidance for stability.
  • Recombinant mouse GDNF fragment corresponding to residues S78-I211.
  • C-terminal 6xHis tag for detection and purification.
  • Produced in E. coli as an animal-free recombinant protein.
  • High purity (≥98.0%) for consistent experimental results.
  • Stable at -20°C; after reconstitution stable at 4°C for 1 week or -20°C for long-term (with carrier protein).
  • Available in small research quantities (2 μg, 10 μg, 50 μg).