Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Granulocyte-macrophage colony-stimulating factor (GM-CSF), porcine, recombinant, His-tagged | >95.0% | 10UG
SDP

Catalog No. 5000177792
Encompass_Preferred

Recombinant, animal-free porcine granulocyte-macrophage colony-stimulating factor (GM-CSF) expressed in E. coli with an N-terminal 6xHis tag. Supplied lyophilized from a PBS/trehalose solution, the protein is purified (>95% by reducing SDS-PAGE) and has a predicted molecular mass of 15.3 kDa (≈17 kDa by SDS-PAGE). Intended for research use in cell culture, bioassays, and biochemical assays; store and handle according to the manufacturer's datasheet.

  • Recombinant porcine GM-CSF, animal-free.
  • N-terminal 6xHis tag for detection and purification.
  • Expressed in E. coli expression system.
  • Lyophilized formulation in PBS with trehalose.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Predicted molecular mass 15.3 kDa (≈17 kDa by SDS-PAGE).
  • Stable when stored at -20°C; reconstitute and aliquot per datasheet.
  • Available in microgram sizes for small-scale research use.

Catalog No. 50-001-77792 Supplier Medchemexpress LLC Supplier No. HYP700239AF10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant, animal-free porcine granulocyte-macrophage colony-stimulating factor (GM-CSF) expressed in E. coli with an N-terminal 6xHis tag. Supplied lyophilized from a PBS/trehalose solution, the protein is purified (>95% by reducing SDS-PAGE) and has a predicted molecular mass of 15.3 kDa (≈17 kDa by SDS-PAGE). Intended for research use in cell culture, bioassays, and biochemical assays; store and handle according to the manufacturer's datasheet.

  • Recombinant porcine GM-CSF, animal-free.
  • N-terminal 6xHis tag for detection and purification.
  • Expressed in E. coli expression system.
  • Lyophilized formulation in PBS with trehalose.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Predicted molecular mass 15.3 kDa (≈17 kDa by SDS-PAGE).
  • Stable when stored at -20°C; reconstitute and aliquot per datasheet.
  • Available in microgram sizes for small-scale research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.