Learn More
Medchemexpress LLC Animal-free GM-CSF protein, mouse (His) | ≥98.0% | 50UG
SDP
Supplier:  Medchemexpress LLC HYP700180AF50UG
Encompass_Preferred
Recombinant mouse granulocyte-macrophage colony-stimulating factor (GM-CSF) with an N-terminal 6xHis tag, produced in an animal-free expression system and supplied as a purified, biologically active protein for in vitro research applications. The product is quality-tested for purity, endotoxin, and biological activity to support cell stimulation and assay use.
  • Produced in an animal-free expression system for reduced variability in cell assays.
  • High purity (≥98.0%) suitable for biochemical and cell-based studies.
  • Low endotoxin (<0.1 EU/μg) to minimize nonspecific immune activation.
  • Demonstrated biological activity (ED50 ~42.2 pg/mL; specific activity >2x10^5 IU/mg).
  • N-terminal 6xHis tag enables detection and affinity purification if needed.
  • Stable when stored frozen; follow supplier storage and handling recommendations.
Catalog No. 50-001-77732