Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Animal-free GRO-beta/CXCL2 protein, human (His) | >95.0% | 10UG
SDP

Catalog No. 5000177647
Encompass_Preferred

Recombinant human GRO-beta (CXCL2) is an animal-free, His-tagged chemokine provided lyophilized for research use. It corresponds to residues A35-N107 and is expressed in E. coli. The preparation is formulated to enhance stability for cell-based assays and biochemical studies and should be aliquoted and stored frozen to maintain activity.

  • Recombinant human CXCL2 (GRO-beta) chemokine corresponding to residues A35-N107.
  • N-terminal 6xHis tag to facilitate purification and detection.
  • Expressed in E. coli and produced without animal-derived materials.
  • Lyophilized formulation from PBS (pH 7.4) with trehalose for improved stability.
  • Purity greater than 95% by reducing SDS-PAGE; observed molecular weight ≈12 kDa.
  • Store frozen; aliquot at -20°C or -80°C to preserve activity; for research use only.

Catalog No. 50-001-77647 Supplier Medchemexpress LLC Supplier No. HYP700045AF10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human GRO-beta (CXCL2) is an animal-free, His-tagged chemokine provided lyophilized for research use. It corresponds to residues A35-N107 and is expressed in E. coli. The preparation is formulated to enhance stability for cell-based assays and biochemical studies and should be aliquoted and stored frozen to maintain activity.

  • Recombinant human CXCL2 (GRO-beta) chemokine corresponding to residues A35-N107.
  • N-terminal 6xHis tag to facilitate purification and detection.
  • Expressed in E. coli and produced without animal-derived materials.
  • Lyophilized formulation from PBS (pH 7.4) with trehalose for improved stability.
  • Purity greater than 95% by reducing SDS-PAGE; observed molecular weight ≈12 kDa.
  • Store frozen; aliquot at -20°C or -80°C to preserve activity; for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.