Learn More
Medchemexpress LLC Interferon beta (human) recombinant, C-terminal 6xHis tag | >95.0% | 10UG
SDP
Catalog No. 5000178331
Encompass_Preferred
Recombinant human interferon-beta (mature sequence M22-N187) with a C-terminal 6xHis tag, expressed in Escherichia coli and supplied as a lyophilized powder for research and cell culture use. The protein is suitable for bioactivity assays and cell-based studies, with low endotoxin and high specific activity.
Catalog No. 50-001-78331 Supplier Medchemexpress LLC Supplier No. HYP700090AF10UG
Recombinant human interferon-beta (mature sequence M22-N187) with a C-terminal 6xHis tag, expressed in Escherichia coli and supplied as a lyophilized powder for research and cell culture use. The protein is suitable for bioactivity assays and cell-based studies, with low endotoxin and high specific activity.