Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Animal-free IFN-lambda 2 (interleukin-28A) protein, mouse, C-terminal His tag | >95.0% | 10UG
SDP

Catalog No. 5000177359
Encompass_Preferred

Animal-Free IFN-lambda 2 (interleukin-28A) is a recombinant mouse cytokine produced in E. coli and supplied with a C-terminal 6xHis tag for research use. The lyophilized protein is intended for cell culture studies and antiviral assays; the manufacturer reports a predicted molecular mass of 20.6 kDa, purity >95% by reducing SDS-PAGE, and biological activity with an ED50 <2 ng/mL. Store at -20°C; after reconstitution it is stable short term at 4°C.

  • Produced in an animal-free process to reduce variability.
  • Expressed in E. coli with a C-terminal 6xHis tag for detection and purification.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • Predicted molecular mass ~20.6 kDa; SDS-PAGE observed 17-25 kDa.
  • Demonstrated biological activity (ED50 <2 ng/mL) in antiviral assays.
  • Recommended storage at -20°C; stable short term at 4°C after reconstitution.

Catalog No. 50-001-77359 Supplier Medchemexpress LLC Supplier No. HYP700203AF10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Animal-Free IFN-lambda 2 (interleukin-28A) is a recombinant mouse cytokine produced in E. coli and supplied with a C-terminal 6xHis tag for research use. The lyophilized protein is intended for cell culture studies and antiviral assays; the manufacturer reports a predicted molecular mass of 20.6 kDa, purity >95% by reducing SDS-PAGE, and biological activity with an ED50 <2 ng/mL. Store at -20°C; after reconstitution it is stable short term at 4°C.

  • Produced in an animal-free process to reduce variability.
  • Expressed in E. coli with a C-terminal 6xHis tag for detection and purification.
  • High purity (>95%) confirmed by reducing SDS-PAGE.
  • Predicted molecular mass ~20.6 kDa; SDS-PAGE observed 17-25 kDa.
  • Demonstrated biological activity (ED50 <2 ng/mL) in antiviral assays.
  • Recommended storage at -20°C; stable short term at 4°C after reconstitution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.